5-Phenylisoxazole-3-carbonyl chloride

Lieferant: Apollo Scientific

OR346228-1G
APOSOR346228-1GEA 0 EUR
APOSOR346228-1G
5-Phenylisoxazole-3-carbonyl chloride
5-Phenylisoxazole-3-carbonyl chloride
Formel: C₁₀H₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07772806
CAS-Nummer: 78189-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR