5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: Ethyl 5-hydroxy-1H-pyrazole-3-carboxylate

OR346230-1G
APOSOR346230-1GEA 42.2 EUR
APOSOR346230-1G
5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester
5-Hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester
Formel: C₆H₈N₂O₃
Molecular Weight: 156.139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263987
CAS-Nummer: 85230-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR