4,6-Dimethyl-pyrimidine-5-carboxylic acid methyl ester
Lieferant: Apollo Scientific
Synonyme:
Methyl 4,6-dimethylpyrimidine-5-carboxylate
OR346240-1G
APOSOR346240-1GEA
217
EUR
APOSOR346240-1G
4,6-Dimethyl-pyrimidine-5-carboxylic acid methyl ester
4,6-Dimethyl-pyrimidine-5-carboxylic acid methyl ester
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166.177 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11046670 CAS-Nummer: 832090-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...