2-tert-Butoxycarbonylamino-4-methyl-oxazole-5-carboxylic acid methyl ester

Lieferant: Apollo Scientific

OR346241-1G
APOSOR346241-1GEA 0 EUR
APOSOR346241-1G
2-tert-Butoxycarbonylamino-4-methyl-oxazole-5-carboxylic acid methyl ester
2-tert-Butoxycarbonylamino-4-methyl-oxazole-5-carboxylic acid methyl ester
Formel: C₁₁H₁₆N₂O₅
Molecular Weight: 256,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19981434
CAS-Nummer: 1311278-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR