2-Amino-6-brom-5-methoxypyridin
Lieferant: Apollo Scientific
OR346242-1G
OR346242-500MG
APOSOR346242-1GEA
369
EUR
APOSOR346242-1G
APOSOR346242-500MG
2-Amino-6-brom-5-methoxypyridin
2-Amino-6-bromo-5-methoxypyridine
Formel:
C₆H₇BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06662268 CAS-Nummer: 79491-43-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...