Benzofuran-3-carboxylic acid methyl ester
Lieferant: Apollo Scientific
|
Achtung
|
OR346248-1G
OR346248-500MG
OR346248-5G
APOSOR346248-1GEA
0
EUR
APOSOR346248-1G
APOSOR346248-500MG
APOSOR346248-5G
Benzofuran-3-carboxylic acid methyl ester
Benzofuran-3-carboxylic acid methyl ester
Formel:
C₁₀H₈O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD15144245 CAS-Nummer: 4687-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...