1-Benzofuran-2-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR346249-1G
OR346249-500MG
APOSOR346249-1GEA
353
EUR
APOSOR346249-1G
APOSOR346249-500MG
1-Benzofuran-2-sulphonyl chloride
1-Benzofuran-2-sulphonyl chloride
Formel:
C₈H₅ClO₃S Schmelzpunkt: 79…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974038 CAS-Nummer: 17070-58-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...