2-Methylpropanethioamide

Lieferant: Apollo Scientific

OR346256-1G
APOSOR346256-1GEA 0 EUR
APOSOR346256-1G
2-Methylpropanethioamide
2-Methylpropanethioamide
Formel: C₄H₉NS
Lagertemperatur: Raumtemperatur
MDL: MFCD07369538
CAS-Nummer: 13515-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR