5-(5-Bromopyridin-2-yl)-3-methylisoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR346260-1G
APOSOR346260-1GEA 0 EUR
APOSOR346260-1G
5-(5-Bromopyridin-2-yl)-3-methylisoxazole-4-carboxylic acid
5-(5-Bromopyridin-2-yl)-3-methylisoxazole-4-carboxylic acid
Formel: C₁₀H₇BrN₂O₃
Molecular Weight: 283,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384844
CAS-Nummer: 1299607-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR