(2Z,4E)-5-Amino-2-cyano-3-methylpenta-2,4-dienoic acid ethyl ester

Lieferant: Apollo Scientific

OR346265-1G
APOSOR346265-1GEA 0 EUR
APOSOR346265-1G
(2Z,4E)-5-Amino-2-cyano-3-methylpenta-2,4-dienoic acid ethyl ester
(2Z,4E)-5-Amino-2-cyano-3-methylpenta-2,4-dienoic acid ethyl ester
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384836
CAS-Nummer: 1300019-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR