5-Nitro-3H-benzofuran-2-one
Lieferant: Apollo Scientific
Synonyme:
5-Nitrobenzofuran-2(3H)-one
OR346267-1G
APOSOR346267-1GEA
369
EUR
APOSOR346267-1G
5-Nitro-3H-benzofuran-2-one
5-Nitro-3H-benzofuran-2-one
Formel:
C₈H₅NO₄ Molecular Weight: 179,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071597 CAS-Nummer: 21997-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...