3-(5-Methyl[1,3,4]oxadiazol-2-yl)phenylamine

Lieferant: Apollo Scientific

OR346268-1G
APOSOR346268-1GEA 0 EUR
APOSOR346268-1G
3-(5-Methyl[1,3,4]oxadiazol-2-yl)phenylamine
3-(5-Methyl[1,3,4]oxadiazol-2-yl)phenylamine
Formel: C₉H₉N₃O
Molecular Weight: 175,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052289
CAS-Nummer: 122733-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR