3-(6-Methylpyridin-2-yl)propan-1-ol

Lieferant: Apollo Scientific

OR346269-1G
APOSOR346269-1GEA 0 EUR
APOSOR346269-1G
3-(6-Methylpyridin-2-yl)propan-1-ol
3-(6-Methylpyridin-2-yl)propan-1-ol
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012344
CAS-Nummer: 61744-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR