(2E,4E)-2-Cyano-5-phenyl-penta-2,4-dienoic acid methyl ester
Lieferant: Apollo Scientific
OR346272-1G
APOSOR346272-1GEA
0
EUR
APOSOR346272-1G
(2E,4E)-2-Cyano-5-phenyl-penta-2,4-dienoic acid methyl ester
(2E,4E)-2-Cyano-5-phenyl-penta-2,4-dienoic acid methyl ester
Formel:
C₁₃H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00999317 CAS-Nummer: 41109-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...