(E)-1-Dimethylamino-4,4-dimethoxy-pent-1-en-3-one

Lieferant: Apollo Scientific

OR346274-1G
APOSOR346274-1GEA 0 EUR
APOSOR346274-1G
(E)-1-Dimethylamino-4,4-dimethoxy-pent-1-en-3-one
(E)-1-Dimethylamino-4,4-dimethoxy-pent-1-en-3-one
Formel: C₉H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18384851
CAS-Nummer: 106157-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR