2,5-Dimethyl-6-oxo-1,6-dihydropyrimidine-4-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR346275-1G
APOSOR346275-1GEA 0 EUR
APOSOR346275-1G
2,5-Dimethyl-6-oxo-1,6-dihydropyrimidine-4-carboxylic acid ethyl ester
2,5-Dimethyl-6-oxo-1,6-dihydropyrimidine-4-carboxylic acid ethyl ester
Formel: C₉H₁₂N₂O₃
Molecular Weight: 196,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00833126
CAS-Nummer: 74536-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR