2,6-Dichloroisonicotinic acid ethyl ester

Lieferant: Apollo Scientific

Synonyme: 2,6-Dichloro-isonicotinic acid ethyl ester, Ethyl 2,6-dichloroisonicotinate

OR346276-1G
APOSOR346276-1GEA 318 EUR
APOSOR346276-1G
2,6-Dichloroisonicotinic acid ethyl ester
2,6-Dichloroisonicotinic acid ethyl ester
Formel: C₈H₇Cl₂NO₂
Molecular Weight: 220,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00235122
CAS-Nummer: 1604-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR