5-Thien-3-ylcyclohexane-1,3-dione

Lieferant: Apollo Scientific

OR346279-1G
APOSOR346279-1GEA 0 EUR
APOSOR346279-1G
5-Thien-3-ylcyclohexane-1,3-dione
5-Thien-3-ylcyclohexane-1,3-dione
Formel: C₁₀H₁₀O₂S
Molecular Weight: 194,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827893
CAS-Nummer: 239132-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR