2-(3-Methoxyphenyl)ethanimidamide hydrochloride

Lieferant: Apollo Scientific

OR346284-1G
APOSOR346284-1GEA 0 EUR
APOSOR346284-1G
2-(3-Methoxyphenyl)ethanimidamide hydrochloride
2-(3-Methoxyphenyl)ethanimidamide hydrochloride
Formel: C₉H₁₃ClN₂O
Molecular Weight: 200,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06761713
CAS-Nummer: 6487-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR