2-(4-Chloro-phenyl)-acetamidine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR346285-1G
APOSOR346285-1GEA 0 EUR
APOSOR346285-1G
2-(4-Chloro-phenyl)-acetamidine hydrochloride
2-(4-Chloro-phenyl)-acetamidine hydrochloride
Formel: C₈H₉ClN₂·ClH
Molecular Weight: 205,084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06761714
CAS-Nummer: 55154-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR