2-Bromo-4-methyl-benzoyl chloride

Lieferant: Apollo Scientific

OR346286-1G
APOSOR346286-1GEA 0 EUR
APOSOR346286-1G
2-Bromo-4-methyl-benzoyl chloride
2-Bromo-4-methyl-benzoyl chloride
Formel: C₈H₆BrClO
Lagertemperatur: Raumtemperatur
MDL: MFCD12025347
CAS-Nummer: 53456-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR