5-Pyridin-2-yl-thiophene-2-sulphonic acid amide

Lieferant: Apollo Scientific

OR346287-1G
APOSOR346287-1GEA 0 EUR
APOSOR346287-1G
5-Pyridin-2-yl-thiophene-2-sulphonic acid amide
5-Pyridin-2-yl-thiophene-2-sulphonic acid amide
Formel: C₉H₈N₂O₂S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00107905
CAS-Nummer: 190659-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR