2-(3,5-Dichlorophenyl)acetonitrile

Lieferant: Apollo Scientific
Achtung

OR346288-1G
APOSOR346288-1GEA 21.9 EUR
APOSOR346288-1G
2-(3,5-Dichlorophenyl)acetonitrile
2-(3,5-Dichlorophenyl)acetonitrile
Formel: C₈H₅Cl₂N
Molecular Weight: 186,0380 g/mol
Schmelzpunkt: 139…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09923674
CAS-Nummer: 52516-37-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR