4-Thiophen-3-yl-but-3-en-2-one

Lieferant: Apollo Scientific

OR346289-1G
APOSOR346289-1GEA 0 EUR
APOSOR346289-1G
4-Thiophen-3-yl-but-3-en-2-one
4-Thiophen-3-yl-but-3-en-2-one
Formel: C₈H₈OS
Molecular Weight: 152,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20286588
CAS-Nummer: 123293-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR