2H-Chromene-3-carbonitrile

Lieferant: Apollo Scientific

OR346291-1G
APOSOR346291-1GEA 0 EUR
APOSOR346291-1G
2H-Chromene-3-carbonitrile
2H-Chromene-3-carbonitrile
Formel: C₁₀H₇NO
Molecular Weight: 157,169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00202019
CAS-Nummer: 57543-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR