3-Methyl-isothiazole-5-sulphonic acid amide
Lieferant: Apollo Scientific
|
Achtung
|
OR346293-100MG
APOSOR346293-100MGEA
0
EUR
APOSOR346293-100MG
3-Methyl-isothiazole-5-sulphonic acid amide
3-Methyl-isothiazole-5-sulphonic acid amide
Formel:
C₄H₆N₂O₂S₂ Molecular Weight: 178,233 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11036155 CAS-Nummer: 1022128-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...