4-(4-Nitro-phenyl)-pyrimidine-2-thiol
Lieferant: Apollo Scientific
OR346298-1G
APOSOR346298-1GEA
0
EUR
APOSOR346298-1G
4-(4-Nitro-phenyl)-pyrimidine-2-thiol
4-(4-Nitro-phenyl)-pyrimidine-2-thiol
Formel:
C₁₀H₇N₃O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD01940450 CAS-Nummer: 155957-44-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...