3-Chloro-4-(propane-2-sulphonyl)-thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

OR346300-1G
APOSOR346300-1GEA 0 EUR
APOSOR346300-1G
3-Chloro-4-(propane-2-sulphonyl)-thiophene-2-carboxylic acid
3-Chloro-4-(propane-2-sulphonyl)-thiophene-2-carboxylic acid
Formel: C₈H₉ClO₄S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00084961
CAS-Nummer: 175202-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR