1-Chloro-2-isopropylsulphanyl-3-nitro-benzene

Lieferant: Apollo Scientific

OR346304-1G
APOSOR346304-1GEA 0 EUR
APOSOR346304-1G
1-Chloro-2-isopropylsulphanyl-3-nitro-benzene
1-Chloro-2-isopropylsulphanyl-3-nitro-benzene
Formel: C₉H₁₀ClNO₂S
Molecular Weight: 231,699 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01312757
CAS-Nummer: 178551-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR