2-Amino-5-tert-butyl-benzonitrile

Lieferant: Apollo Scientific

OR346307-1G
APOSOR346307-1GEA 306 EUR
APOSOR346307-1G
2-Amino-5-tert-butyl-benzonitrile
2-Amino-5-tert-butyl-benzonitrile
Formel: C₁₁H₁₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06408810
CAS-Nummer: 874814-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR