2-Chloropyrimidine-5-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Pyrimidinesulphonyl chloride, 2-chloro-

OR346309-1G
APOSOR346309-1GEA 0 EUR
APOSOR346309-1G
2-Chloropyrimidine-5-sulphonyl chloride
2-Chloropyrimidine-5-sulphonyl chloride
Formel: C₄H₂Cl₂N₂O₂S
Molecular Weight: 213,04 g/mol
Schmelzpunkt: 64…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06657208
CAS-Nummer: 98026-88-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR