Diethyl 2-oxoheptyl phosphonate

Lieferant: Apollo Scientific

OR346314-1G
APOSOR346314-1GEA 0 EUR
APOSOR346314-1G
Diethyl 2-oxoheptyl phosphonate
Diethyl 2-oxoheptyl phosphonate
Formel: C₁₁H₂₃O₄P
Molecular Weight: 250,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573488
CAS-Nummer: 3450-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR