3-(3-Methyl-isothiazol-5-yl)-benzaldehyde

Lieferant: Apollo Scientific

OR346317-500MG
APOSOR346317-500MGEA 340 EUR
APOSOR346317-500MG
3-(3-Methyl-isothiazol-5-yl)-benzaldehyde
3-(3-Methyl-isothiazol-5-yl)-benzaldehyde
Formel: C₁₁H₉NOS
Molecular Weight: 203.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573495
CAS-Nummer: 1401522-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR