2-(3-Methyl-isothiazol-5-yl)-benzaldehyde

Lieferant: Apollo Scientific

OR346318-500MG
APOSOR346318-500MGEA 0 EUR
APOSOR346318-500MG
2-(3-Methyl-isothiazol-5-yl)-benzaldehyde
2-(3-Methyl-isothiazol-5-yl)-benzaldehyde
Formel: C₁₁H₉NOS
Molecular Weight: 203,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573496
CAS-Nummer: 1401521-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR