Imidazo[2,1-b]thiazol-6-ylmethanol
Lieferant: Apollo Scientific
OR346325-1G
APOSOR346325-1GEA
0
EUR
APOSOR346325-1G
Imidazo[2,1-b]thiazol-6-ylmethanol
Imidazo[2,1-b]thiazol-6-ylmethanol
Formel:
C₆H₆N₂OS Molecular Weight: 154,190 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368518 CAS-Nummer: 349480-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...