Methyl-6-(hydroxymethyl)nicotinat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Hydroxymethyl)pyridine-3-carboxylate methyll ester, Methyl 6-(hydroxymethyl)nicotinate, 6-(Hydroxymethyl)nicotinic acid methyll ester, Methyl 6-(Hydroxymethyl)pyridine-3-carboxylate

OR346326-1G
APOSOR346326-1GEA 122 EUR
APOSOR346326-1G
Methyl-6-(hydroxymethyl)nicotinat 95%
Methyl-6-(hydroxymethyl)nicotinate
Formel: C₈H₉NO₃
Schmelzpunkt: 77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00643821
CAS-Nummer: 56026-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR