4-Methoxy-benzaldehyde oxime

Lieferant: Apollo Scientific

OR346329-1G OR346329-5G
APOSOR346329-1GEA 72.5 EUR
APOSOR346329-1G APOSOR346329-5G
4-Methoxy-benzaldehyde oxime
4-Methoxy-benzaldehyde oxime
Formel: C₈H₉NO₂
Molecular Weight: 151.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00137433
CAS-Nummer: 3235-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR