(E)-3-(2-Cyano-phenyl)-acrylic acid
Lieferant: Apollo Scientific
OR346331-1G
APOSOR346331-1GEA
0
EUR
APOSOR346331-1G
(E)-3-(2-Cyano-phenyl)-acrylic acid
(E)-3-(2-Cyano-phenyl)-acrylic acid
Formel:
C₁₀H₇NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00189382 CAS-Nummer: 2886-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...