(3-Oxo-2,3-dihydro-1H-isoindol-1-yl)-acetic acid

Lieferant: Apollo Scientific

OR346333-1G
APOSOR346333-1GEA 0 EUR
APOSOR346333-1G
(3-Oxo-2,3-dihydro-1H-isoindol-1-yl)-acetic acid
(3-Oxo-2,3-dihydro-1H-isoindol-1-yl)-acetic acid
Formel: C₁₀H₉NO₃
Molecular Weight: 191.183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00456083
CAS-Nummer: 3849-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR