2-Mercapto-7,8-dihydro-5H-thiopyrano[4,3-d]pyrimidin-4-ol

Lieferant: Apollo Scientific

OR346334-1G
APOSOR346334-1GEA 0 EUR
APOSOR346334-1G
2-Mercapto-7,8-dihydro-5H-thiopyrano[4,3-d]pyrimidin-4-ol
2-Mercapto-7,8-dihydro-5H-thiopyrano[4,3-d]pyrimidin-4-ol
Formel: C₇H₈N₂OS₂
Molecular Weight: 200,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573501
CAS-Nummer: 150787-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR