(3-Oxo-2,3-dihydro-1H-isoindol-1-yl)-acetic acid methyl ester

Lieferant: Apollo Scientific

OR346336-1G
APOSOR346336-1GEA 0 EUR
APOSOR346336-1G
(3-Oxo-2,3-dihydro-1H-isoindol-1-yl)-acetic acid methyl ester
(3-Oxo-2,3-dihydro-1H-isoindol-1-yl)-acetic acid methyl ester
Formel: C₁₁H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18417615
CAS-Nummer: 28488-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR