4-(5-Methyl-[1,2,4]oxadiazol-3-yl)-benzoic acid methyl ester

Lieferant: Apollo Scientific

OR346337-1G
APOSOR346337-1GEA 0 EUR
APOSOR346337-1G
4-(5-Methyl-[1,2,4]oxadiazol-3-yl)-benzoic acid methyl ester
4-(5-Methyl-[1,2,4]oxadiazol-3-yl)-benzoic acid methyl ester
Formel: C₁₁H₁₀N₂O₃
Molecular Weight: 218,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198877
CAS-Nummer: 196301-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR