2-(4-tert-Butyl-benzyl)-isothiourea hydrochloride

Lieferant: Apollo Scientific

OR346338-1G
APOSOR346338-1GEA 0 EUR
APOSOR346338-1G
2-(4-tert-Butyl-benzyl)-isothiourea hydrochloride
2-(4-tert-Butyl-benzyl)-isothiourea hydrochloride
Formel: C₁₂H₁₈N₂S·ClH
Molecular Weight: 258.811 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10677023
CAS-Nummer: 324523-97-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR