3,5-Dihydroxy-[1,2,4]triazine-6-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR346340-1G
APOSOR346340-1GEA
142
EUR
APOSOR346340-1G
3,5-Dihydroxy-[1,2,4]triazine-6-carboxylic acid
3,5-Dihydroxy-[1,2,4]triazine-6-carboxylic acid
Formel:
C₄H₃N₃O₄ Molecular Weight: 157,084 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20542230 CAS-Nummer: 13924-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...