1-Methyl-3-(2-nitro-phenyl)-1H-pyrazole

Lieferant: Apollo Scientific

OR346341-1G
APOSOR346341-1GEA 0 EUR
APOSOR346341-1G
1-Methyl-3-(2-nitro-phenyl)-1H-pyrazole
1-Methyl-3-(2-nitro-phenyl)-1H-pyrazole
Formel: C₁₀H₉N₃O₂
Molecular Weight: 203,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573508
CAS-Nummer: 305811-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR