4-Isopropyl-3-thiosemicarbazid

Lieferant: Apollo Scientific

Synonyme: N-Isopropylhydrazinecarbothioamide

OR346343-1G
APOSOR346343-1GEA 0 EUR
APOSOR346343-1G
4-Isopropyl-3-thiosemicarbazid
4-Isopropyl-3-thiosemicarbazid
Formel: C₄H₁₁N₃S
Molecular Weight: 133,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025144
CAS-Nummer: 13431-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR