2-Cyano-3-(3,4-dichloro-phenyl)-acrylic acid ethyl ester

Lieferant: Apollo Scientific

OR346344-1G
APOSOR346344-1GEA 0 EUR
APOSOR346344-1G
2-Cyano-3-(3,4-dichloro-phenyl)-acrylic acid ethyl ester
2-Cyano-3-(3,4-dichloro-phenyl)-acrylic acid ethyl ester
Formel: C₁₂H₉Cl₂NO₂
Molecular Weight: 270,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00087072
CAS-Nummer: 55417-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR