2-Bromo-N-phenyl-acetamide

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-N-phenylacetamide

OR346345-1G
APOSOR346345-1GEA 0 EUR
APOSOR346345-1G
2-Bromo-N-phenyl-acetamide
2-Bromo-N-phenyl-acetamide
Formel: C₈H₈BrNO
Molecular Weight: 214,059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00462518
CAS-Nummer: 5326-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR