1-Methyl-2,3-dihydro-1H-indole

Lieferant: Apollo Scientific
Achtung

OR346348-1G
APOSOR346348-1GEA 0 EUR
APOSOR346348-1G
1-Methyl-2,3-dihydro-1H-indole
1-Methyl-2,3-dihydro-1H-indole
Formel: C₉H₁₁N
Lagertemperatur: Raumtemperatur
MDL: MFCD01718890
CAS-Nummer: 88475-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR