(5-Ethylsulphanyl-[1,3,4]oxadiazol-2-ylmethyl)-carbamic acid tert-butyl ester
Lieferant: Apollo Scientific
OR346349-1G
APOSOR346349-1GEA
0
EUR
APOSOR346349-1G
(5-Ethylsulphanyl-[1,3,4]oxadiazol-2-ylmethyl)-carbamic acid tert-butyl ester
(5-Ethylsulphanyl-[1,3,4]oxadiazol-2-ylmethyl)-carbamic acid tert-butyl ester
Formel:
C₁₀H₁₇N₃O₃S Molecular Weight: 259,325 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00436038 CAS-Nummer: 914216-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...